siRNA Details
Gene Symbol: CCDC192 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317938.1 | NP_001304867.1 | 4, 5 | 416 | ||
XM_017009805.1 | XP_016865294.1 | 3, 4 | 478 | |||
XM_017009806.1 | XP_016865295.1 | 4, 5 | 382 | |||
XM_017009807.1 | XP_016865296.1 | 4, 5 | 373 | |||
XM_017009808.1 | XP_016865297.1 | 3, 4 | 304 | |||
XM_017009809.1 | XP_016865298.1 | 4, 5 | 447 | |||
XM_017009810.1 | XP_016865299.1 | 4, 5 | 446 | |||
XM_017009811.1 | XP_016865300.1 | 4, 5 | 446 | |||
XM_017009812.1 | XP_016865301.1 | 4, 5 | 447 | |||
XM_017009813.1 | XP_016865302.1 | 4, 5 | 445 | |||
GenBank mRNA | HY052603.1 | - | 4, 5 | 384 |
