siRNA Details
Gene Symbol: FLJ46906 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC104460.2 | - | 1 | 211 | ||
BC104461.2 | - | 1 | 211 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: FLJ46906 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC104460.2 | - | 1 | 211 | ||
BC104461.2 | - | 1 | 211 |