siRNA Details
Gene Symbol: VIMP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018445.5 | NP_060915.2 | 6 | 1203 | ||
GenBank mRNA | AF157317.1 | - | 6 | 1129 | ||
AF328864.1 | - | 6 | 1130 | |||
AK026455.1 | - | 6 | 1166 | |||
AK097011.1 | - | 4 | 2393 | |||
AK225955.1 | - | 6 | 1129 | |||
AY324824.1 | - | 6 | 1130 | |||
BC005840.2 | - | 6 | 1126 | 2967406 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
