siRNA Details
Gene Symbol: MIR124-2HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034102.2 | 2 | 721 | |||
NR_034103.2 | 2 | 542 | ||||
NR_109792.1 | 2 | 611 | ||||
NR_109793.1 | 2 | 583 | ||||
GenBank mRNA | AK124256.1 | - | 2 | 612 | ||
BX537900.1 | - | 1 | 503 |
