siRNA Details
Gene Symbol: TTTY7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003592.1 | 5 | 855 | |||
GenBank mRNA | BX089948.1 | - | 2 | 353 |
Gene Symbol: TTTY7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_001534.2 | 5 | 855 | |||
GenBank mRNA | AF278762.1 | - | 2 | 453 |
