siRNA Details
Gene Symbol: LOC101927021 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110065.1 | 4 | 911 | |||
NR_110066.1 | 3 | 593 | ||||
NR_110067.1 | 4 | 849 | ||||
NR_110068.1 | 2 | 549 | ||||
NR_110069.1 | 3 | 741 | ||||
NR_110070.1 | 5 | 700 | ||||
GenBank mRNA | AK090593.1 | - | 3 | 475 | ||
BM544796.1 | - | 5 | 721 | |||
DA025114.1 | - | 3 | 475 | |||
DA689880.1 | - | 2 | 429 |
