siRNA Details
Gene Symbol: LOC101927021 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110065.1 | 4 | 1542 | |||
NR_110066.1 | 3 | 1224 | ||||
NR_110067.1 | 4 | 1480 | ||||
NR_110068.1 | 2 | 1180 | ||||
NR_110069.1 | 3 | 1372 | ||||
NR_110070.1 | 5 | 1331 | ||||
GenBank mRNA | AK090593.1 | - | 3 | 1106 |
