siRNA Details
Gene Symbol: GS1-259H13.2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195541.1 | NP_001182470.1 | 4 | 503 | ||
NM_001195542.1 | NP_001182471.1 | 4 | 500 | |||
NM_001195543.1 | NP_001182472.1 | 4 | 494 | |||
XM_005250088.4 | XP_005250145.1 | 4 | 420 | |||
XM_011515691.2 | XP_011513993.1 | 5 | 640 | |||
XM_011515692.2 | XP_011513994.1 | 5 | 637 | |||
XM_011515694.2 | XP_011513996.1 | 5 | 628 | |||
XM_011515695.2 | XP_011513997.1 | 5 | 631 | |||
XM_011515696.2 | XP_011513998.1 | 5 | 578 | |||
XM_011515697.1 | XP_011513999.1 | 5 | 520 | |||
XM_011515699.2 | XP_011514001.1 | 4 | 472 | |||
XM_011515700.2 | XP_011514002.1 | 4 | 459 | |||
XM_017011629.1 | XP_016867118.1 | 5 | 563 | |||
GenBank mRNA | BG461486.1 | - | 2 | 322 | ||
BI559535.1 | - | 4 | 497 | |||
DB054606.1 | - | 4 | 494 |
