siRNA Details
Gene Symbol: GS1-259H13.2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195541.1 | NP_001182470.1 | 6 | 866 | ||
NM_001195542.1 | NP_001182471.1 | 6 | 863 | |||
NM_001195543.1 | NP_001182472.1 | 6 | 857 | |||
XM_005250088.4 | XP_005250145.1 | 6 | 783 | |||
XM_011515691.2 | XP_011513993.1 | 7 | 1003 | |||
XM_011515692.2 | XP_011513994.1 | 7 | 1000 | |||
XM_011515694.2 | XP_011513996.1 | 7 | 991 | |||
XM_011515695.2 | XP_011513997.1 | 7 | 994 | |||
XM_011515696.2 | XP_011513998.1 | 7 | 941 | |||
XM_011515697.1 | XP_011513999.1 | 7 | 883 | |||
XM_011515699.2 | XP_011514001.1 | 6 | 835 | |||
XM_011515700.2 | XP_011514002.1 | 6 | 822 | |||
XM_017011629.1 | XP_016867118.1 | 7 | 926 |
