siRNA Details
Gene Symbol: GS1-259H13.2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195541.1 | NP_001182470.1 | 3 | 203 | ||
NM_001195542.1 | NP_001182471.1 | 3 | 200 | |||
NM_001195543.1 | NP_001182472.1 | 3 | 194 | |||
XM_005250088.4 | XP_005250145.1 | 3 | 120 | |||
XM_011515691.2 | XP_011513993.1 | 4 | 340 | |||
XM_011515692.2 | XP_011513994.1 | 4 | 337 | |||
XM_011515694.2 | XP_011513996.1 | 4 | 328 | |||
XM_011515695.2 | XP_011513997.1 | 4 | 331 | |||
XM_011515696.2 | XP_011513998.1 | 4 | 278 | |||
XM_011515697.1 | XP_011513999.1 | 4 | 220 | |||
XM_011515699.2 | XP_011514001.1 | 3 | 172 | |||
XM_011515700.2 | XP_011514002.1 | 3 | 159 | |||
XM_017011629.1 | XP_016867118.1 | 4 | 263 | |||
GenBank mRNA | BG461486.1 | - | 1 | 22 | ||
BI559535.1 | - | 3 | 197 | |||
DB054606.1 | - | 3 | 194 | |||
DB442398.1 | - | 3 | 203 | |||
DB456868.1 | - | 3 | 194 | |||
DB462723.1 | - | 3 | 178 |
