siRNA Details
Gene Symbol: ZNF181 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029997.3 | NP_001025168.2 | 4 | 1009 | ||
NM_001145665.1 | NP_001139137.1 | 4 | 1006 | |||
XM_005258849.2 | XP_005258906.1 | 5 | 777 | |||
XM_005258850.2 | XP_005258907.1 | 5 | 774 | |||
XM_005258851.3 | XP_005258908.1 | 5 | 765 | |||
XM_006723183.3 | XP_006723246.1 | 5 | 1031 | |||
XM_006723184.3 | XP_006723247.1 | 5 | 762 | |||
XM_017026739.1 | XP_016882228.1 | 5 | 1519 | |||
XM_017026740.1 | XP_016882229.1 | 4 | 639 | |||
GenBank mRNA | AK295492.1 | - | 5 | 676 | ||
BC043228.1 | - | 3 | 1650 | 5295602 | ||
BC104759.1 | - | 4 | 635 | 8143762 | ||
BC143442.1 | - | 4 | 632 | 9051950 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
