siRNA Details
Gene Symbol: ZNF853 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017560.1 | NP_060030.1 | 3 | 3153 | ||
XM_011515438.2 | XP_011513740.1 | 4 | 3315 | |||
XM_011515439.2 | XP_011513741.1 | 4 | 3319 | |||
XM_017012371.1 | XP_016867860.1 | 3 | 3433 | |||
GenBank mRNA | AL390175.1 | - | 1 | 331 |
