siRNA Details
Gene Symbol: KIAA1210 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020721.1 | NP_065772.1 | 4 | 626 | ||
XM_006724774.3 | XP_006724837.1 | 3 | 323 | |||
XM_017029688.1 | XP_016885177.1 | 3 | 362 | |||
XM_017029689.1 | XP_016885178.1 | 3 | 362 | |||
GenBank mRNA | DV080213.1 | - | 1 | 133 |
