siRNA Details
Gene Symbol: KIAA1257 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB033083.1 | - | 8 | 4436 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: KIAA1257 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB033083.1 | - | 8 | 4436 |