siRNA Details
Gene Symbol: EBF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110514.1 | NP_001103984.1 | 3 | 418 | ||
XM_006723600.1 | XP_006723663.1 | 3 | 213 | |||
XM_017027983.1 | XP_016883472.1 | 4 | 310 | |||
XM_017027984.1 | XP_016883473.1 | 4 | 416 | |||
GenBank mRNA | AB037863.1 | - | 3 | 412 |
