siRNA Details
Gene Symbol: EBF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110514.1 | NP_001103984.1 | 16, 17 | 2004 | ||
XM_006723600.1 | XP_006723663.1 | 16, 17 | 1799 | |||
XM_017027983.1 | XP_016883472.1 | 17, 18 | 1896 | |||
XM_017027984.1 | XP_016883473.1 | 17, 18 | 2002 | |||
XM_017027985.1 | XP_016883474.1 | 10, 11 | 1158 | |||
GenBank mRNA | AB037863.1 | - | 16, 17 | 1882 | ||
BC019106.2 | - | 2, 3 | 495 | |||
BC037555.1 | - | 8, 9 | 1104 | |||
BC054347.1 | - | 1, 2 | 658 |
