siRNA Details
Gene Symbol: BAHCC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291324.1 | NP_001278253.1 | 14 | 4547 | ||
XM_011525063.2 | XP_011523365.1 | 15 | 4939 | |||
XM_017024898.1 | XP_016880387.1 | 14 | 4598 | |||
XM_017024899.1 | XP_016880388.1 | 13 | 4504 | |||
GenBank mRNA | AB040880.1 | - | 10 | 2298 |
