siRNA Details
Gene Symbol: TRIM36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300759.1 | NP_001287688.1 | 2 | 412 | ||
NM_018700.3 | NP_061170.2 | 2 | 628 | |||
XM_017009621.1 | XP_016865110.1 | 2 | 589 | |||
XM_017009622.1 | XP_016865111.1 | 2 | 1306 | |||
XM_017009623.1 | XP_016865112.1 | 2 | 589 | |||
GenBank mRNA | AJ272269.1 | - | 2 | 245 | ||
AK296389.1 | - | 2 | 410 | |||
AK316422.1 | - | 2 | 315 | |||
BC130334.1 | - | 2 | 297 | 40146365 | ||
DC397736.1 | - | 2 | 414 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
