siRNA Details
Gene Symbol: LOC645166 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027355.2 | 4 | 405 | |||
NR_027356.2 | 3 | 327 | ||||
GenBank mRNA | AW954772.1 | - | 3 | 316 | ||
BQ438821.1 | - | 4 | 406 | |||
CR746216.1 | - | 4 | 432 | |||
DB496177.1 | - | 3 | 314 |
Gene Symbol: LOC107985911 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017005465.1 | XP_016860954.1 | 4 | 889 |
Gene Symbol: LOC654342 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027238.1 | 4 | 373 | |||
GenBank mRNA | BF031534.1 | - | 4 | 373 | ||
BM826054.1 | - | 3 | 232 |
