siRNA Details
Gene Symbol: ADGB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024694.3 | NP_078970.3 | 3 | 298 | ||
XM_006715566.3 | XP_006715629.1 | 3 | 333 | |||
XM_011536125.2 | XP_011534427.1 | 3 | 284 | |||
XM_011536126.1 | XP_011534428.1 | 3 | 333 | |||
XM_011536127.2 | XP_011534429.1 | 3 | 333 | |||
XM_011536128.2 | XP_011534430.1 | 3 | 333 | |||
XM_011536130.2 | XP_011534432.1 | 4 | 232 | |||
XM_011536134.2 | XP_011534436.1 | 3 | 293 | |||
XM_017011314.1 | XP_016866803.1 | 3 | 333 | |||
XM_017011315.1 | XP_016866804.1 | 3 | 333 | |||
GenBank mRNA | AK097570.1 | - | 3 | 298 | ||
AL832192.1 | - | 3 | 295 |
