siRNA Details
Gene Symbol: CAMSAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015447.3 | NP_056262.3 | 7, 8 | 1040 | ||
XM_005263396.3 | XP_005263453.1 | 8, 9 | 1465 | |||
XM_005263397.1 | XP_005263454.1 | 5, 6 | 566 | |||
XM_011518272.1 | XP_011516574.1 | 7, 8 | 734 | |||
XM_011518273.2 | XP_011516575.1 | 6, 7 | 565 | |||
XM_011518274.1 | XP_011516576.1 | 5, 6 | 595 | |||
XM_017014301.1 | XP_016869790.1 | 7, 8 | 1321 | |||
XM_017014302.1 | XP_016869791.1 | 7, 8 | 1245 | |||
GenBank mRNA | AF116701.2 | - | 7, 8 | 1019 | ||
AJ519841.1 | - | 5, 6 | 565 | |||
BC130580.1 | - | 7, 8 | 699 | 40146611 | ||
BC144082.1 | - | 7, 8 | 699 | 9052602 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
