siRNA Details
Gene Symbol: ANGPTL8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018687.6 | NP_061157.3 | 2, 3 | 477 | ||
GenBank mRNA | AF271350.2 | - | 4, 5 | 828 | ||
AY358356.1 | - | 2, 3 | 461 | |||
BC113823.1 | - | 2, 3 | 234 | |||
BP316755.1 | - | 2, 3 | 478 | |||
KF809856.1 | - | 2, 3 | 395 | |||
KF809857.1 | - | 2, 3 | 249 |
