siRNA Details
Gene Symbol: LOC100128554 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130748.1 | 7 | 1276 | |||
GenBank mRNA | BC035235.1 | - | 8 | 1452 | ||
BC039370.1 | - | 7 | 1276 | |||
BC118583.1 | - | 7 | 820 | 40085030 | ||
BC121801.1 | - | 8 | 1059 |
