siRNA Details
Gene Symbol: LANCL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018697.3 | NP_061167.1 | 2, 3 | 908 | ||
GenBank mRNA | AB035966.1 | - | 2, 3 | 385 | ||
AF353942.1 | - | 2, 3 | 385 | |||
AJ278245.1 | - | 2, 3 | 516 | |||
AK095857.1 | - | 2, 3 | 908 | |||
AK301747.1 | - | 2, 3 | 314 | |||
AK313326.1 | - | 2, 3 | 653 | |||
AL713665.2 | - | 2, 3 | 300 | |||
BC070049.1 | - | 2, 3 | 604 | 30343188 |
