siRNA Details
Gene Symbol: TRIM36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300759.1 | NP_001287688.1 | 2, 3 | 599 | ||
NM_018700.3 | NP_061170.2 | 2, 3 | 815 | |||
GenBank mRNA | AJ272269.1 | - | 2, 3 | 432 | ||
AK296389.1 | - | 2, 3 | 597 | |||
AK316422.1 | - | 2, 3 | 502 | |||
BC130334.1 | - | 2, 3 | 484 | 40146365 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
