siRNA Details
Gene Symbol: FAR2P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046258.1 | 6 | 2006 | |||
NR_046259.1 | 5 | 1908 | ||||
NR_046260.1 | 4 | 1783 | ||||
GenBank mRNA | AK054990.1 | - | 4 | 1223 | ||
BC131487.1 | - | 2 | 3831 |
Gene Symbol: FAR2P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026758.2 | 12 | 3682 |
