siRNA Details
Gene Symbol: LOC101927196 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110214.1 | 2 | 249 | |||
NR_110215.1 | 2 | 207 | ||||
NR_110216.1 | 2 | 212 | ||||
GenBank mRNA | BC039382.1 | - | 2 | 205 | ||
BQ429883.1 | - | 2 | 182 | |||
HY037094.1 | - | 2 | 250 | |||
HY052589.1 | - | 2 | 238 |
