siRNA Details
Gene Symbol: SLCO4C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_180991.4 | NP_851322.3 | 13 | 4701 | ||
XM_011543370.1 | XP_011541672.1 | 12 | 4379 | |||
XM_011543372.1 | XP_011541674.1 | 15 | 4352 | |||
GenBank mRNA | AK128854.1 | - | 13 | 4425 | ||
BX647349.1 | - | 13 | 4590 | |||
BX647577.1 | - | 2 | 2522 | |||
BX647880.1 | - | 2 | 2522 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
