siRNA Details
Gene Symbol: TTTY14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_125734.1 | 3 | 629 | |||
NR_125735.1 | 2 | 554 | ||||
NR_125736.1 | 4 | 304 | ||||
GenBank mRNA | AF119903.1 | - | LIT2233 CYorf14 |
1 | 1071 | |
BX280014.1 | - | 2 | 280 | |||
FY211103.1 | - | 2 | 217 | |||
FY211594.1 | - | 3 | 293 | |||
HY121109.1 | - | 4 | 297 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
