siRNA Details
Gene Symbol: ZNF836 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102657.1 | NP_001096127.1 | 5 | 796 | ||
XM_011526558.2 | XP_011524860.1 | 5 | 638 | |||
XM_011526559.2 | XP_011524861.1 | 4 | 683 | |||
XM_017026389.1 | XP_016881878.1 | 5 | 571 | |||
GenBank mRNA | AK131308.1 | - | 5 | 648 |
