siRNA Details
Gene Symbol: ZNF846 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077624.1 | NP_001071092.1 | 6 | 1071 | ||
XM_005259772.3 | XP_005259829.1 | 6 | 862 | |||
XM_005259773.4 | XP_005259830.1 | 6 | 1068 | |||
XM_005259775.3 | XP_005259832.1 | 6 | 859 | |||
XM_006722658.3 | XP_006722721.1 | 6 | 1071 | |||
XM_011527717.2 | XP_011526019.1 | 6 | 874 | |||
XM_011527718.2 | XP_011526020.1 | 6 | 871 | |||
XM_011527719.2 | XP_011526021.1 | 6 | 890 | |||
XM_011527720.2 | XP_011526022.1 | 5 | 645 | |||
XM_011527721.2 | XP_011526023.1 | 4 | 419 | |||
XM_017026404.1 | XP_016881893.1 | 6 | 1029 | |||
XM_017026405.1 | XP_016881894.1 | 5 | 1511 | |||
XM_017026406.1 | XP_016881895.1 | 3 | 349 | |||
XM_017026407.1 | XP_016881896.1 | 6 | 1071 | |||
XM_017026408.1 | XP_016881897.1 | 6 | 762 | |||
GenBank mRNA | AK097652.1 | - | 5 | 1164 | ||
AK289536.1 | - | 2 | 672 | |||
AK308938.1 | - | 5 | 871 | |||
BC118576.1 | - | 6 | 642 | 40053684 | ||
BC118657.1 | - | 5 | 559 |
