siRNA Details
Gene Symbol: PARL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324436.1 | NP_001311365.1 | 6, 7 | 827 | ||
NM_001324438.1 | NP_001311367.1 | 6, 7 | 827 | |||
NM_018622.6 | NP_061092.3 | 6, 7 | 827 | |||
NR_136893.1 | 6, 7 | 883 | ||||
XM_005247582.4 | XP_005247639.1 | 6, 7 | 822 | |||
XM_017006800.1 | XP_016862289.1 | 6, 7 | 822 | |||
GenBank mRNA | AF197937.1 | - | 6, 7 | 765 | ||
AK024765.1 | - | 6, 7 | 799 | |||
BC014058.2 | - | 6, 7 | 785 | 3937238 |
Gene Symbol: AIG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017010927.1 | XP_016866416.1 | 6 | 1196 | ||
XM_017010928.1 | XP_016866417.1 | 5 | 1062 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
