siRNA Details
Gene Symbol: C20orf194 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009984.2 | NP_001009984.1 | 20, 21 | 1791 | ||
XM_005260684.3 | XP_005260741.1 | 20, 21 | 1785 | |||
XM_011529208.2 | XP_011527510.1 | 1, 2 | 128 | |||
XM_017027783.1 | XP_016883272.1 | 17, 18 | 1412 | |||
GenBank mRNA | AK023919.1 | - | 2, 3 | 453 | ||
AK226100.1 | - | 15, 16 | 1215 | |||
AK303375.1 | - | 11, 12 | 1160 | |||
AL110249.1 | - | 11, 12 | 756 | |||
BC122529.1 | - | 11, 12 | 1009 | 40012183 |
