siRNA Details
Gene Symbol: C20orf194 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009984.2 | NP_001009984.1 | 25 | 2309 | ||
XM_005260684.3 | XP_005260741.1 | 25 | 2303 | |||
XM_005260687.4 | XP_005260744.1 | 6 | 1145 | |||
XM_011529208.2 | XP_011527510.1 | 6 | 646 | |||
XM_017027783.1 | XP_016883272.1 | 22 | 1930 | |||
GenBank mRNA | AK226100.1 | - | 20 | 1733 | ||
AK303375.1 | - | 16 | 1678 | |||
AL110249.1 | - | 16 | 1274 | |||
BC122529.1 | - | 16 | 1527 | 40012183 |
