siRNA Details
Gene Symbol: GOLGA6A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038640.2 | NP_001033729.2 | 17, 18 | 1999 | ||
GenBank mRNA | AF263742.1 | - | 17, 18 | 1999 | ||
AK292574.1 | - | 17, 18 | 1980 |
Gene Symbol: GOLGA6D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145224.1 | NP_001138696.1 | 17, 18 | 1993 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
