siRNA Details
Gene Symbol: DISC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012957.1 | NP_001012975.1 | 13 | 2836 | ||
NM_001164537.1 | NP_001158009.1 | 14 | 2998 | |||
NM_001164540.1 | NP_001158012.1 | 11 | 2536 | |||
NM_018662.2 | NP_061132.2 | 13 | 2902 | |||
GenBank mRNA | AB007926.1 | - | 13 | 2822 | ||
AF222980.1 | - | 13 | 2902 | |||
BC151225.1 | - | 13 | 2822 | 9007180 | ||
FJ804174.1 | - | 12 | 2766 | |||
FJ804175.1 | - | 12 | 2751 | |||
FJ804176.1 | - | 12 | 2847 | |||
FJ804177.1 | - | 11 | 2536 | |||
FJ804178.1 | - | 12 | 2799 | |||
FJ804179.1 | - | 14 | 2998 | |||
FJ804180.1 | - | 13 | 2894 |
Gene Symbol: TSNAX-DISC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028393.1 | 16 | 3515 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
