siRNA Details
Gene Symbol: LINC00639 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_039982.1 | 5 | 4162 | |||
GenBank mRNA | AK125018.1 | - | 7 | 4352 | ||
AK127318.1 | - | 6 | 3519 | |||
BC035119.1 | - | 6 | 4229 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00639 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_039982.1 | 5 | 4162 | |||
GenBank mRNA | AK125018.1 | - | 7 | 4352 | ||
AK127318.1 | - | 6 | 3519 | |||
BC035119.1 | - | 6 | 4229 |