siRNA Details
Gene Symbol: ZNF302 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF155656.1 | - | 5 | 2186 | ||
AF265236.1 | - | 6 | 2522 | |||
AF326206.1 | - | 5 | 2285 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
