siRNA Details
Gene Symbol: THSD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018676.3 | NP_061146.1 | 3 | 1452 | ||
NM_199263.2 | NP_954872.1 | 3 | 1452 | |||
GenBank mRNA | AB044385.1 | - | 3 | 1058 | ||
AK096289.1 | - | 3 | 1452 | |||
AK315087.1 | - | 3 | 1101 | |||
AY358149.1 | - | 3 | 1115 | |||
BC063842.1 | - | 3 | 1085 | 6181100 |
