siRNA Details
Gene Symbol: FRG2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001124759.1 | NP_001118231.1 | 4 | 1187 | ||
XM_011533254.2 | XP_011531556.1 | 5 | 2020 | |||
XM_011533256.2 | XP_011531558.1 | 5 | 2017 | |||
XM_017005489.1 | XP_016860978.1 | 5 | 2097 |
