siRNA Details
Gene Symbol: TRIM36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300752.1 | NP_001287681.1 | 7 | 1608 | ||
NM_001300759.1 | NP_001287688.1 | 8 | 1627 | |||
NM_018700.3 | NP_061170.2 | 8 | 1843 | |||
XM_017009621.1 | XP_016865110.1 | 10 | 1855 | |||
XM_017009622.1 | XP_016865111.1 | 10 | 2572 | |||
XM_017009623.1 | XP_016865112.1 | 10 | 1855 | |||
GenBank mRNA | AB196558.1 | - | 7 | 1099 | ||
AJ272269.1 | - | 8 | 1460 | |||
AK058179.1 | - | 3 | 484 | |||
AK296389.1 | - | 8 | 1625 | |||
AK316422.1 | - | 8 | 1530 | |||
BC130334.1 | - | 8 | 1512 | 40146365 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
