siRNA Details
Gene Symbol: LINC01096 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015450.1 | 1 | 945 | |||
GenBank mRNA | AK058104.1 | - | 2 | 459 | ||
BC048110.1 | - | 1 | 922 | |||
BC053955.1 | - | 1 | 922 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC01096 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015450.1 | 1 | 945 | |||
GenBank mRNA | AK058104.1 | - | 2 | 459 | ||
BC048110.1 | - | 1 | 922 | |||
BC053955.1 | - | 1 | 922 |