siRNA Details
Gene Symbol: LOC105375650 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_132986.1 | 2 | 764 | |||
NR_132987.1 | 2 | 714 |
Gene Symbol: LOC286149 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK093190.1 | - | 2 | 714 |
