siRNA Details
Gene Symbol: MROH2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287395.1 | NP_001274324.1 | 33 | 3800 | ||
XM_011511075.2 | XP_011509377.1 | 33 | 4022 | |||
XM_011511076.2 | XP_011509378.1 | 33 | 4022 | |||
XM_011511078.2 | XP_011509380.1 | 33 | 3983 | |||
XM_011511079.2 | XP_011509381.1 | 33 | 3977 | |||
XM_011511080.2 | XP_011509382.1 | 33 | 5493 | |||
XM_011511081.2 | XP_011509383.1 | 32 | 3917 | |||
XM_011511082.2 | XP_011509384.1 | 33 | 4022 | |||
XM_011511083.2 | XP_011509385.1 | 33 | 4022 | |||
XM_011511084.2 | XP_011509386.1 | 32 | 3824 | |||
XM_011511085.2 | XP_011509387.1 | 27 | 3092 |
