siRNA Details
Gene Symbol: LINC00298 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015405.1 | 3 | 444 | |||
GenBank mRNA | BC043361.1 | - | 4 | 515 | ||
BC103700.1 | - | 3 | 444 | |||
BC104747.1 | - | 4 | 562 | |||
BC105688.1 | - | 3 | 444 | |||
BC107115.1 | - | 3 | 444 |
