siRNA Details
Gene Symbol: ZNF720 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK128671.1 | - | 2 | 1097 | ||
AK299345.1 | - | 6 | 874 | |||
AL832197.1 | - | 5 | 794 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ZNF720 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK128671.1 | - | 2 | 1097 | ||
AK299345.1 | - | 6 | 874 | |||
AL832197.1 | - | 5 | 794 |