siRNA Details
Gene Symbol: ZNF284 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037813.2 | NP_001032902.1 | 5 | 564 | ||
XM_011526907.2 | XP_011525209.1 | 5 | 605 | |||
XM_011526908.2 | XP_011525210.1 | 6 | 665 | |||
GenBank mRNA | AK308295.1 | - | 11 | 2273 | ||
AY166789.1 | - | 3 | 266 | |||
AY458845.1 | - | 5 | 564 | |||
CR936662.1 | - | 5 | 507 |
