siRNA Details
Gene Symbol: MCIDAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190787.1 | NP_001177716.1 | 4 | 547 | ||
XM_017009439.1 | XP_016864928.1 | 2 | 208 | |||
GenBank mRNA | FR854393.1 | - | 4 | 370 |
