siRNA Details
Gene Symbol: TOPAZ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145030.1 | NP_001138502.1 | 2 | 1194 | ||
XM_011533694.2 | XP_011531996.1 | 2 | 1194 | |||
XM_017006361.1 | XP_016861850.1 | 2 | 1194 | |||
XM_017006362.1 | XP_016861851.1 | 2 | 1194 | |||
GenBank mRNA | AK093476.1 | - | 2 | 1194 |
