siRNA Details
Gene Symbol: AQP7P5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017015366.1 | XP_016870855.1 | 9 | 1773 | ||
XM_017015367.1 | XP_016870856.1 | 8 | 1690 | |||
XM_017015368.1 | XP_016870857.1 | 8 | 1673 | |||
XM_017015369.1 | XP_016870858.1 | 9 | 1756 | |||
XM_017015370.1 | XP_016870859.1 | 5 | 1773 | |||
XM_017015371.1 | XP_016870860.1 | 7 | 1540 | |||
XM_017015373.1 | XP_016870862.1 | 6 | 1411 | |||
XM_017015374.1 | XP_016870863.1 | 6 | 1349 | |||
XM_017015375.1 | XP_016870864.1 | 5 | 1265 |
Gene Symbol: AQP7P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002817.2 | 4 | 1666 | |||
GenBank mRNA | AK128231.1 | - | 4 | 1662 | ||
BC070322.1 | - | 4 | 989 |
